Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acad12
Entrez Gene ID
338350
Gene Name
acyl-Coenzyme A dehydrogenase family, member 12
Gene Aliases
9330129D05Rik
Chromosome Location
Chr.5: 121598277 - 121618938 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178799.3 NP_848914.2 9 - 10 1616 75
GenBank mRNA AK033963.1 - 9 - 10 1535 75

View Full Product Details