Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rrp15
Entrez Gene ID
67223
Gene Name
ribosomal RNA processing 15 homolog (S. cerevisiae)
Gene Aliases
2810430M08Rik, 5430437H21Rik
Chromosome Location
Chr.1: 186721086 - 186749358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026041.2 NP_080317.3 4 - 5 751 138
GenBank mRNA AK007616.1 - 4 - 5 705 138
AK010658.1 - 4 - 5 719 138
AK013217.1 - 4 - 5 719 138
AK087997.1 - 4 - 5 721 138
AK169088.1 - 4 - 5 752 138
BC006787.1 - 4 - 5 717 138
CO043575.1 - 1 - 2 138

View Full Product Details