The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007552.4 | NP_031578.2 | 6 - 7 | 901 | 150 |
GenBank mRNA | BC056384.1 | - | 5 - 6 | 431 | 150 |
DQ337175.1 | - | 5 - 6 | 430 | 150 | |
M64279.1 | - | 6 - 7 | 901 | 150 |