The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024206.4 | NP_077168.2 | 4 - 5 | 351 | 116 |
GenBank mRNA | AK003354.1 | - | 4 - 5 | 352 | 116 |
AK009755.1 | - | 3 - 4 | 335 | 116 |