The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025403.4 | NP_079679.1 | 1 - 2 | 145 | 85 |
GenBank mRNA | AK004120.1 | - | 1 - 2 | 146 | 85 |
AK012875.1 | - | 1 - 2 | 102 | 85 | |
BC028497.1 | - | 1 - 2 | 100 | 85 |