Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Commd2
Entrez Gene ID
52245
Gene Name
COMM domain containing 2
Gene Aliases
1190017G07Rik, AI047739, D3Ertd176e, HSPC042
Chromosome Location
Chr.3: 57644349 - 57651684 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175095.4 NP_780304.2 5 - 5 2242 77
GenBank mRNA AK083784.1 - 5 - 5 2250 77
AK143821.1 - 5 - 5 2243 77
BC062098.1 - - 1971 77

View Full Product Details