The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026160.4 | NP_080436.1 | 4 - 4 | 1157 | 141 |
GenBank mRNA | AK003106.1 | - | 4 - 4 | 1156 | 141 |
AK132329.1 | - | - | 1166 | 141 | |
AK172322.1 | - | 1 - 1 | 716 | 141 | |
BC068180.1 | - | 4 - 4 | 1160 | 141 | |
BC086774.1 | - | 3 - 3 | 3584 | 141 | |
BC110308.1 | - | 4 - 4 | 1146 | 141 |