The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013840.3 | NP_038868.2 | 7 - 7 | 765 | 90 |
GenBank mRNA | AK027917.1 | - | 7 - 7 | 766 | 90 |