The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020618.4 | NP_065643.1 | 11 - 11 | 2430 | 140 |
GenBank mRNA | AK018493.1 | - | 11 - 11 | 1859 | 139 |
AK076177.1 | - | 11 - 11 | 1868 | 140 | |
BC047141.1 | - | 11 - 11 | 1852 | 140 | |
BC061498.1 | - | 11 - 11 | 1861 | 140 | |
BC065043.1 | - | 11 - 11 | 2430 | 140 |