Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Stx1b
Entrez Gene ID
56216
Gene Name
syntaxin 1B
Gene Aliases
Stx1b2, Stx1bl, Syn1b
Chromosome Location
Chr.7: 127803900 - 127824531 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024414.2 NP_077725.1 3 - 3 420 153
GenBank mRNA BC132042.1 - 3 - 3 215 153
BC132068.1 - 3 - 3 215 153
D29743.1 - 3 - 3 247 153

View Full Product Details