The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033954.3 | NP_001029126.1 | 1 - 2 | 147 | 84 |
NM_007587.2 | NP_031613.2 | 1 - 2 | 147 | 84 | |
GenBank mRNA | AF330212.1 | - | 1 - 2 | 85 | 84 |
AK132582.1 | - | 1 - 2 | 97 | 84 | |
BC028771.1 | - | 1 - 2 | 80 | 84 | |
BC030071.1 | - | 1 - 2 | 78 | 84 | |
CJ060164.1 | - | 1 - 2 | 148 | 84 | |
X97991.1 | - | 1 - 2 | 108 | 84 |