The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007587.2 | NP_031613.2 | 3 - 4 | 384 | 73 |
GenBank mRNA | AK132582.1 | - | 3 - 4 | 334 | 73 |
BC030071.1 | - | 3 - 4 | 315 | 73 | |
X97991.1 | - | 3 - 4 | 345 | 73 |