Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cr2
Entrez Gene ID
12902
Gene Name
complement receptor 2
Gene Aliases
C3DR, CD21, CD35, Cr-1, Cr-2, Cr1
Chromosome Location
Chr.1: 195136811 - 195177190 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007758.2 NP_031784.1 5 - 6 920 120
GenBank mRNA AK003072.2 - 5 - 6 920 120
BC109344.1 - 5 - 6 853 120
M35684.1 - 4 - 5 788 120
M61132.1 - 4 - 5 755 120

View Full Product Details