The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008277.2 | NP_032303.1 | 6 - 7 | 418 | 72 |
GenBank mRNA | AK149416.1 | - | 7 - 8 | 456 | 72 |
BC013343.1 | - | 6 - 7 | 442 | 72 | |
D29987.1 | - | 7 - 8 | 456 | 72 | |
X59530.1 | - | 6 - 7 | 408 | 72 |