Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nxph2
Entrez Gene ID
18232
Gene Name
neurexophilin 2
Gene Aliases
-
Chromosome Location
Chr.2: 23320899 - 23401986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008752.2 NP_032778.1 1 - 2 292 91
GenBank mRNA AK038402.1 - 1 - 2 293 91
BC106863.1 - 1 - 2 262 91
BC106864.2 - 1 - 2 262 91

View Full Product Details