Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plxna2
Entrez Gene ID
18845
Gene Name
plexin A2
Gene Aliases
2810428A13Rik, AA589422, AW457381, OCT, PlexA2, Plxn2, mKIAA0463
Chromosome Location
Chr.1: 194619829 - 194816869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008882.2 NP_032908.2 6 - 7 2373 71
GenBank mRNA AK122289.1 - 5 - 6 637 71
BC068155.1 - 6 - 7 2155 71
D86949.1 - 6 - 7 2279 71

View Full Product Details