Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptgir
Entrez Gene ID
19222
Gene Name
prostaglandin I receptor (IP)
Gene Aliases
IP, PGI2
Chromosome Location
Chr.7: 16905419 - 16910905 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK033567.1 - 1 - 2 429 66

View Full Product Details