The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009300.3 | NP_033326.1 | 2 - 3 | 398 | 70 |
GenBank mRNA | AK020349.1 | - | 2 - 3 | 399 | 70 |
M35732.1 | - | 2 - 3 | 363 | 70 |