The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253885.1 | NP_001240814.1 | 5 - 6 | 1414 | 69 |
NM_001253886.1 | NP_001240815.1 | 5 - 6 | 1414 | 69 | |
NM_001253887.1 | NP_001240816.1 | 5 - 6 | 414 | 69 | |
NM_009685.3 | NP_033815.1 | 5 - 6 | 1228 | 69 | |
GenBank mRNA | AK030748.1 | - | 5 - 6 | 1111 | 69 |
AK075730.1 | - | 5 - 6 | 394 | 69 | |
AK083830.1 | - | 5 - 6 | 1144 | 69 | |
AK144340.1 | - | 5 - 6 | 1415 | 69 | |
AK164140.1 | - | 5 - 6 | 1139 | 69 | |
BC036956.1 | - | 4 - 5 | 1052 | 69 | |
BC048395.1 | - | 3 - 4 | 560 | 69 |