The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201413.1 | NP_001188342.1 | 14 - 15 | 2383 | 93 |
NM_001201414.1 | NP_001188343.1 | 13 - 14 | 2386 | 93 | |
NM_001201415.1 | NP_001188344.1 | 13 - 14 | 2320 | 93 | |
NM_001201416.1 | NP_001188345.1 | 4 - 5 | 486 | 93 | |
NM_009686.2 | NP_033816.1 | 15 - 16 | 2389 | 93 | |
GenBank mRNA | AK004792.1 | - | 15 - 16 | 2243 | 93 |
AK153260.1 | - | 13 - 14 | 2352 | 93 | |
AK167649.1 | - | 14 - 15 | 2248 | 93 | |
AK169959.1 | - | 13 - 14 | 2258 | 93 | |
AK172405.1 | - | 4 - 5 | 490 | 93 | |
BC042797.1 | - | 5 - 6 | 528 | 93 | |
BC076587.1 | - | 13 - 14 | 2166 | 93 |