Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aqp4
Entrez Gene ID
11829
Gene Name
aquaporin 4
Gene Aliases
WCH4
Chromosome Location
Chr.18: 15389394 - 15410977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009700.2 NP_033830.2 4 - 5 747 69
GenBank mRNA AF469168.1 - 4 - 5 821 69
AF469169.1 - 4 - 5 1003 69
AK039777.1 - 4 - 5 750 69
AK045357.1 - 3 - 4 398 69
AY484964.1 - 4 - 5 753 69
AY484965.1 - 5 - 6 937 69
AY484966.1 - 5 - 6 883 69
AY728043.1 - 4 - 5 767 69
AY728044.1 - 5 - 6 951 69
AY728045.1 - 4 - 5 879 69
AY803942.1 - 5 - 6 858 69
AY803943.1 - 6 - 7 988 69
AY803944.1 - 7 - 8 1172 69
BC024526.1 - 4 - 5 693 69
CT010362.1 - 3 - 4 694 69
U33012.1 - 3 - 4 709 69
U48397.1 - 3 - 4 829 69
U48398.1 - 3 - 4 700 69
U48399.1 - 5 - 6 811 69
U88623.1 - 4 - 5 748 69

View Full Product Details