The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243052.1 | NP_001229981.1 | 4 - 5 | 665 | 66 |
NM_001243053.1 | NP_001229982.1 | 5 - 6 | 550 | 66 | |
NM_009737.3 | NP_033867.1 | 4 - 5 | 638 | 66 | |
GenBank mRNA | AF031467.1 | - | 4 - 5 | 430 | 66 |
AK037482.1 | - | 4 - 5 | 443 | 66 | |
AK137049.1 | - | 5 - 6 | 601 | 66 | |
AK145300.1 | - | 4 - 5 | 483 | 66 | |
AK160159.1 | - | 4 - 5 | 666 | 66 | |
BB630667.1 | - | 4 - 5 | 441 | 66 | |
BC017688.1 | - | 4 - 5 | 415 | 66 | |
BC048072.1 | - | 4 - 5 | 429 | 66 | |
U68526.1 | - | 4 - 5 | 424 | 66 |