Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myl3
Entrez Gene ID
17897
Gene Name
myosin, light polypeptide 3
Gene Aliases
MLC1s, MLC1v, Mylc, VLC1
Chromosome Location
Chr.9: 110763681 - 110769794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010859.2 NP_034989.1 2 - 3 243 75
GenBank mRNA AK002312.1 - 2 - 3 245 75
AK011518.1 - 2 - 3 236 75
AK146943.1 - 2 - 3 247 75
BC061222.1 - 2 - 3 246 75

View Full Product Details