Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kdm5d
Entrez Gene ID
20592
Gene Name
lysine (K)-specific demethylase 5D
Gene Aliases
HY, Jarid1d, Smcy
Chromosome Location
Chr.Y: 897660 - 943813 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011419.3 NP_035549.1 9 - 10 1417 73
GenBank mRNA AB221607.1 - 9 - 10 1243 73
AF127244.1 - 9 - 10 1255 73
AK047790.1 - 9 - 10 1418 73
AK140971.1 - 9 - 10 1412 73
AK141096.1 - 8 - 9 3870 73
BC059077.1 - 1 - 2 232 73
BC060077.1 - - 1396 73
Z29652.1 - 9 - 10 1173 73

View Full Product Details