The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011879.2 | NP_036009.1 | 19 - 20 | 1767 | 83 |
GenBank mRNA | AK082356.1 | - | 19 - 20 | 1700 | 83 |
AK087976.1 | - | 19 - 20 | 1713 | 83 | |
BC006706.1 | - | 17 - 18 | 2125 | 83 | |
BC014739.1 | - | 19 - 20 | 1698 | 83 | |
BC026569.1 | - | 19 - 20 | 1698 | 83 | |
BC029148.1 | - | 17 - 18 | 2661 | 83 | |
BC029671.1 | - | 19 - 20 | 1710 | 83 | |
BC029694.1 | - | 19 - 20 | 1707 | 83 | |
BC033356.1 | - | 19 - 20 | 1694 | 83 | |
BM211698.2 | - | 1 - 2 | 83 |