Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmp1
Entrez Gene ID
13406
Gene Name
dentin matrix protein 1
Gene Aliases
AV020965, Dmp, PP
Chromosome Location
Chr.5: 104197558 - 104214109 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016779.2 NP_058059.2 4 - 5 253 118
GenBank mRNA AK132177.1 - 4 - 5 254 118
AK157973.1 - 4 - 5 254 118
BC113753.1 - 4 - 5 175 118
U65020.1 - 4 - 5 259 118

View Full Product Details