Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk32b
Entrez Gene ID
64293
Gene Name
serine/threonine kinase 32B
Gene Aliases
2510009F08Rik, STKG6, Stk32, YANK2
Chromosome Location
Chr.5: 37446825 - 37717153 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022416.2 NP_071861.1 11 - 12 1469 87
GenBank mRNA AJ250840.1 - 11 - 12 1547 87
AK082468.1 - 11 - 12 1470 87
BC052404.1 - 11 - 12 1379 87
BC056396.1 - 11 - 12 1455 87
BC058412.1 - 11 - 12 1379 87

View Full Product Details