The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026453.3 | NP_080729.1 | 5 - 6 | 437 | 92 |
GenBank mRNA | AK015369.1 | - | 5 - 6 | 478 | 92 |
AK075982.1 | - | 5 - 6 | 438 | 92 | |
AK075983.1 | - | 5 - 6 | 438 | 92 | |
BC013079.1 | - | 5 - 6 | 452 | 92 |