The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026965.2 | NP_081241.1 | 2 - 3 | 257 | 94 |
GenBank mRNA | AK007659.1 | - | 2 - 3 | 255 | 94 |
AK054334.1 | - | 2 - 3 | 258 | 94 | |
BC019467.1 | - | 2 - 3 | 241 | 94 | |
BC049670.1 | - | 2 - 3 | 252 | 94 | |
CT010388.1 | - | 2 - 3 | 219 | 94 |