Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rusc1
Entrez Gene ID
72296
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
2210403N08Rik, AA408288, NESCA
Chromosome Location
Chr.3: 89083975 - 89093363 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083807.1 NP_001077276.1 3 - 4 1622 82
NM_001083808.1 NP_001077277.1 2 - 3 162 82
NM_028188.2 NP_082464.2 4 - 5 2033 82
GenBank mRNA AK032334.1 - 2 - 3 652 82
AK039664.1 - 4 - 5 1961 82
AK170012.1 - 2 - 3 177 82
AK170210.1 - 2 - 3 125 82
BC052081.1 - 2 - 3 366 82
BC056360.1 - 3 - 4 1560 82
BC057034.1 - 2 - 3 228 82
BY201665.1 - 2 - 3 163 82

View Full Product Details