Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rusc1
Entrez Gene ID
72296
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
2210403N08Rik, AA408288, NESCA
Chromosome Location
Chr.3: 89083975 - 89093363 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083807.1 NP_001077276.1 4 - 5 1699 57
NM_001083808.1 NP_001077277.1 3 - 4 239 57
NM_028188.2 NP_082464.2 5 - 6 2110 57
GenBank mRNA AK032334.1 - 3 - 4 729 57
AK039664.1 - 5 - 6 2038 57
AK042689.1 - 4 - 5 1647 57
AK170012.1 - 3 - 4 254 57
AK170210.1 - 3 - 4 202 57
BC056360.1 - 4 - 5 1637 57
BC057034.1 - 3 - 4 305 57
BY201665.1 - 3 - 4 240 57

View Full Product Details