The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171007.1 | NP_001164478.1 | 8 - 9 | 2750 | 94 |
NM_172729.3 | NP_766317.1 | 8 - 9 | 3001 | 94 | |
GenBank mRNA | AK082663.1 | - | 5 - 6 | 1796 | 94 |
AK089662.1 | - | 8 - 9 | 2751 | 94 | |
AK137585.1 | - | 8 - 9 | 3006 | 94 | |
AY160222.1 | - | 7 - 8 | 2536 | 94 | |
BC042670.1 | - | 8 - 9 | 2828 | 94 | |
BC043670.1 | - | 5 - 6 | 1718 | 94 |