The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182805.2 | NP_877957.1 | 7 - 8 | 1064 | 66 |
GenBank mRNA | AK008086.1 | - | 7 - 8 | 1087 | 66 |
AK009457.1 | - | 7 - 8 | 1087 | 66 | |
BC022625.1 | - | 7 - 8 | 1065 | 66 | |
BC026846.1 | - | 7 - 8 | 1063 | 66 |