Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Raver2
Entrez Gene ID
242570
Gene Name
ribonucleoprotein, PTB-binding 2
Gene Aliases
A430091O22Rik
Chromosome Location
Chr.4: 101069038 - 101152370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183024.1 NP_898845.1 8 - 9 1448 66
GenBank mRNA AK029114.1 - 6 - 7 944 66
BC055329.1 - 8 - 9 1448 66

View Full Product Details