Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet3
Entrez Gene ID
194388
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
B430006D22Rik, BC037432, D230004J03Rik
Chromosome Location
Chr.6: 83362373 - 83457313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183138.2 NP_898961.2 9 - 10 3121 74
GenBank mRNA AK084164.1 - 4 - 5 474 74
HQ423151.1 - 9 - 10 3295 74
HQ423152.1 - 8 - 9 3238 74
JX946278.1 - 10 - 11 3827 74

View Full Product Details