Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcrl5
Entrez Gene ID
329693
Gene Name
Fc receptor-like 5
Gene Aliases
Fcrh3, mBXMH2
Chromosome Location
Chr.3: 87435782 - 87500678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113238.1 NP_001106709.1 4 - 5 731 96
NM_183222.3 NP_899045.3 5 - 6 995 96
GenBank mRNA AK089756.1 - 4 - 5 96
AY158090.1 - 5 - 6 96
AY506558.1 - 5 - 6 96
BC137997.1 - 5 - 6 96
BC137998.1 - 5 - 6 96

View Full Product Details