The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113238.1 | NP_001106709.1 | 7 - 8 | 1403 | 130 |
NM_183222.3 | NP_899045.3 | 8 - 9 | 1667 | 130 | |
GenBank mRNA | AK089756.1 | - | 7 - 8 | 1373 | 130 |
AY158090.1 | - | 8 - 9 | 1664 | 130 | |
AY506558.1 | - | 8 - 9 | 1673 | 130 | |
BC137997.1 | - | 8 - 9 | 1601 | 130 | |
BC137998.1 | - | 8 - 9 | 1601 | 130 |