Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taldo1
Entrez Gene ID
21351
Gene Name
transaldolase 1
Gene Aliases
-
Chromosome Location
Chr.7: 141392160 - 141402976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011528.4 NP_035658.1 7 - 8 1040 103
GenBank mRNA AK003092.1 - 7 - 8 1004 103
AK142030.1 - 7 - 8 998 103
BC003970.1 - 7 - 8 973 103
BC004754.1 - 7 - 8 995 103
BC094277.1 - 7 - 8 989 103
U67611.1 - 7 - 8 983 103

View Full Product Details