The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183141.2 | NP_898964.2 | 2 - 2 | 4805 | 76 |
GenBank mRNA | AK031970.1 | - | 1 - 1 | 4395 | 76 |
AK173294.1 | - | 1 - 1 | 3271 | 76 | |
BC076582.1 | - | 2 - 2 | 5046 | 76 | |
BC079588.1 | - | 1 - 1 | 4305 | 76 | |
BC094219.1 | - | 2 - 2 | 4805 | 76 |