The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022987.3 | NP_075363.1 | 1 - 1 | 1181 | 118 |
GenBank mRNA | AB042155.1 | - | 1 - 1 | 1203 | 118 |
AB114214.1 | - | 1 - 1 | 840 | 118 |