Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsg4
Entrez Gene ID
16769
Gene Name
desmoglein 4
Gene Aliases
CDHF13, lah
Chromosome Location
Chr.18: 20436175 - 20471821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181564.2 NP_853543.1 4 - 5 506 86
GenBank mRNA AB240192.1 - 4 - 5 506 86
AY191584.1 - 4 - 5 506 86
AY227349.1 - 4 - 5 372 86
BC109009.2 - 4 - 5 506 86
BC150475.2 - 4 - 5 506 86

View Full Product Details