Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dsg4
Entrez Gene ID
16769
Gene Name
desmoglein 4
Gene Aliases
CDHF13, lah
Chromosome Location
Chr.18: 20436175 - 20471821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181564.2 NP_853543.1 9 - 10 1404 148
GenBank mRNA AB240192.1 - 8 - 9 1587 148
AY170623.1 - 2 - 3 352 148
AY191584.1 - 9 - 10 1404 148
AY227349.1 - 9 - 10 1270 148
BC109009.2 - 9 - 10 1404 148
BC150475.2 - 9 - 10 1404 148

View Full Product Details