The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183286.2 | NP_899109.2 | 2 - 3 | 305 | 150 |
GenBank mRNA | AK011939.1 | - | 2 - 3 | 280 | 150 |
AK046465.1 | - | 1 - 2 | 352 | 150 | |
BB637905.1 | - | 2 - 3 | 305 | 150 |