Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem59l
Entrez Gene ID
67937
Gene Name
transmembrane protein 59-like
Gene Aliases
5330410G16Rik
Chromosome Location
Chr.8: 70483863 - 70487468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182991.2 NP_892036.1 2 - 3 337 59
GenBank mRNA AK005155.1 - 2 - 3 301 59
BC056172.1 - 2 - 3 337 59

View Full Product Details