The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164559.1 | NP_001158031.1 | 10 - 11 | 1291 | 116 |
NM_001164560.1 | NP_001158032.1 | 10 - 11 | 1215 | 116 | |
NM_198020.2 | NP_932137.2 | 10 - 11 | 1318 | 116 | |
GenBank mRNA | AK032945.1 | - | 8 - 9 | 2751 | 116 |
AK084775.1 | - | 10 - 11 | 1257 | 117 | |
AK155251.1 | - | 10 - 11 | 1284 | 116 | |
AK159467.1 | - | 10 - 11 | 1256 | 116 | |
AK170412.1 | - | 10 - 11 | 1180 | 116 | |
AK171002.1 | - | 10 - 11 | 1283 | 116 | |
AK178569.1 | - | 3 - 4 | 194 | 116 | |
BC027404.1 | - | 10 - 11 | 1287 | 116 |