Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Luzp2
Entrez Gene ID
233271
Gene Name
leucine zipper protein 2
Gene Aliases
9330154K17Rik, D130073E01
Chromosome Location
Chr.7: 54835200 - 55268888 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178705.5 NP_848820.4 11 - 12 1648 98
GenBank mRNA AK032204.1 - 11 - 12 1305 98
AK034077.1 - 11 - 12 1266 98
AK034240.1 - 11 - 12 1300 98
AY164459.1 - 11 - 12 1645 98
BC075646.1 - 11 - 12 1278 98

View Full Product Details