The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285991.1 | NP_001272920.1 | 9 - 10 | 955 | 90 |
NM_001285993.1 | NP_001272922.1 | 9 - 10 | 922 | 90 | |
NM_028162.4 | NP_082438.3 | 9 - 10 | 955 | 90 | |
GenBank mRNA | AK005461.1 | - | 9 - 10 | 939 | 90 |
AK033710.1 | - | 9 - 10 | 847 | 90 | |
AK144467.1 | - | 9 - 10 | 956 | 90 | |
AK154975.1 | - | 9 - 10 | 915 | 90 | |
AK220251.1 | - | 9 - 10 | 922 | 90 | |
BC043113.1 | - | 9 - 10 | 771 | 90 |