The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025650.2 | NP_079926.1 | 3 - 3 | 399 | 68 |
GenBank mRNA | AK003033.1 | - | 3 - 3 | 409 | 68 |
AK003443.1 | - | 3 - 3 | 379 | 68 | |
AK146292.1 | - | 3 - 3 | 541 | 68 | |
AK167946.1 | - | 3 - 3 | 395 | 68 | |
AK168974.1 | - | 3 - 3 | 364 | 68 | |
BC034408.1 | - | 3 - 3 | 363 | 68 | |
BU529273.1 | - | 3 - 3 | 400 | 68 |