The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009225.2 | NP_033251.1 | 3 - 4 | 396 | 200 |
GenBank mRNA | AK077537.1 | - | 3 - 4 | 349 | 201 |
AK146319.1 | - | 3 - 4 | 439 | 200 | |
BG086527.2 | - | 3 - 4 | 354 | 200 |