Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncl
Entrez Gene ID
17975
Gene Name
nucleolin
Gene Aliases
B530004O11Rik, C23, D0Nds28, D1Nds28, Nucl
Chromosome Location
Chr.1: 86344719 - 86359455 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010880.3 NP_035010.3 12 - 13 2054 109
GenBank mRNA AF318184.1 - 12 - 13 1966 109
AK028681.1 - 12 - 13 1964 109
AK029128.1 - 12 - 13 1964 109
AK031606.1 - 12 - 13 2000 109
AK050958.1 - 10 - 11 1351 109
AK083307.1 - 12 - 13 1974 109
AK088402.1 - 12 - 13 1965 109
AK144894.1 - 12 - 13 1964 109
AK161706.1 - 12 - 13 1974 109
AK163275.1 - 12 - 13 1965 109
AK165167.1 - 10 - 11 2646 109
AK166681.1 - 12 - 13 1964 109
AK168579.1 - 12 - 13 1964 109
AK168623.1 - 12 - 13 1964 109
BC005460.1 - 12 - 13 1874 109

View Full Product Details